Learn More
DWK Life Sciences Wheaton™ PP Made Delivery Nozzles
Shop All Wheaton Science Products Products
Click to view available options
Diameter (Metric):
2 and 3 mm
6 mm
8 mm
Unit Size:
Pack of 6
Description
- Glass sinker is used to secure tubing when drawing from a container
Specifications
Specifications
| Diameter (Metric) | 8 mm |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With | Wheaton peristaltic pumps, dispensing pumps and tubing |
| Product Type | Delivery Nozzle |