Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for Lab 45 Graduated Bottles
Click to view available options
Color:
Blue
Red
White
Includes:
Cap without inner sealing ring
PTFE/Silicone Liner
Wedge shaped inner sealing ring
Description
- 45mm screw thread
- Polypropylene
- Autoclavable
- Linerless
Specifications
For Use With (Equipment) | Lab 45 graduated bottle |
Color | Blue |
Includes | Wedge shaped inner sealing ring |
Material | Polypropylene |
No. per Case | 12 Cs. |
Certifications/Compliance | ISO 9001 |
Autoclavable | Autoclavable |