Learn More
DWK Life Sciences Wheaton™ PBT Screw Caps
Product Code. 10572004 Shop All Wheaton Science Products Products
Click to view available options
Thread Size (Metric):
40 mm
Unit Size:
Pack of 10
Product Code. 10572004
Brand: DWK Life Sciences 240750
Only null left
Add to basket
Add to basket
Withstands temperatures up to 200°C (392°F)
  • With PTFE faced silicone liner
  • Closure Material: Polybutylene Terephthalate (PBT)
  • Closure Size: 45mm (1.77 in.)
  • Closure Color: Red
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Color Red
Material PBT
No. per Case 10 Cs.
Thread Size (Metric) 40 mm
Liner Material PTFE/Silicone
Neck Finish 40 mm
Certifications/Compliance ISO 9001
For Use With (Equipment) Bottle