Learn More
DWK Life Sciences Wheaton™ Microlink Caps
Product Code. 11501235 Shop All Wheaton Science Products Products
Change view
Click to view available options
Neck Finish:
33-430
38-430
Color:
Black
White
Product Type:
Open Top Screw Cap
Screw Cap
No. per Case:
24/Cs.
50/Cs.
Unit Size:
Pack of 24
Pack of 50
Product Code. Neck Finish Color Product Type No. per Case unitSize
11501235 33-430 Black Screw Cap 50/Cs. Pack of 50
12403881 38-430 Black Open Top Screw Cap 50/Cs. Pack of 50
12493871 33-430 Black Open Top Screw Cap 50/Cs. Pack of 50
12433911 38-430 White Screw Cap 24/Cs. Pack of 24
4 options
Product Code. 11501235
Brand: DWK Life Sciences 240106
Only null left
Add to basket
Add to basket
Open top caps are ideal for storing and containing volatile solvents, sensitive compounds and corrosive chemicals
  • Polypropylene/silicone/PTFE liner is ultrasonically welded to each cap to help eliminate possibility of toxicity
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Polypropylene
No. per Case 50/Cs.
Liner Material Polypropylene/Silicone
Neck Finish 33-430
Certifications/Compliance ISO 9001
Product Type Screw Cap
For Use With (Equipment) Bottle