Learn More
DWK Life Sciences Wheaton™ Micro Tissue Grinder With Cap, PTFE Pestle and Cap Liner
Product Code. 10377131 Shop All Wheaton Science Products Products
Click to view available options
Capacity:
0.016 oz., 0.5mL
Unit Size:
Pack of 2
Product Code. 10377131
Brand: DWK Life Sciences 357535
Only null left
Add to basket
Add to basket
Black phenolic 13-425 open-top screw cap with PTFE-faced silicone rubber liner reduces aerosoling of sample. Unit can be autoclaved or sterilized to prevent cross contamination between samples.
  • Pestle rod made from No. 316 stainless steel rod with pestle tip made from PTFE
  • Mortar made from low-extractable borosilicate glass
  • Replace open-top screw cap with solid top cap provides vial for temporary sample storage.
Order Info
Clearance between pestle and mortar is 0.05 to 0.15mm. The sample can be stored in mortar by using a Wheaton solid-top black phenolic screw cap with PTFE-faced rubber liner (No. 240408).
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 0.016 oz.
Autoclavable Autoclavable
Length (English) 4.52 in.
Includes Cap, PTFE pestle, cap liner, Borosilicate Glass mortar
Capacity (Metric) 0.5 mL
Length (Metric) 115mm
Capacity 0.016 oz., 0.5mL