Learn More
DWK Life Sciences Wheaton™ Magna-Flex™ Spinner Flasks
Sfoglia Tutto Wheaton Science Products Prodotti
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
3000 mL
6 L
Includes:
Flask only
Flask with impeller
missing translation for 'unitSize'
Each
Descrizione
- Borosilicate glass flasks
- Two large side arms with screw-cap closures
- Flex-type, bulb-shaped glass impeller rotates from a fixed position above the liquid level around an indentation in the base of the flask
- Graduated
- Removable stainless-steel pin immobilizes the impeller during handling or decanting to prevent damage to cells or microcarriers
- Autoclavable flasks and impellers for selected sizes can also be ordered separately
- U.S. Patent No. 3,854,704
Specifica
Specifica
| Diameter (Metric) Side Arm | 4.5 cm |
| Capacity (Metric) | 6 L |
| Material | Borosilicate Glass |
| Includes | Flask with impeller |
| Autoclavable | Autoclavable |
| Graduated | Yes |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto