Learn More
DWK Life Sciences Wheaton™ M-T Vial File™ and Accessories
Shop All Wheaton Science Products Products
Click to view available options
Product Type:
Index Card
M-T Vial File
Partition
Storage Box
Quantity:
35 Case
6 Case
For Use With (Equipment):
228778 Hinged Case
228780 Hinged Case
24 x 20mL vials
40 x 4mL vials
54 x 8mL vials
60 x 2mL vials
W228790 Hinged Case
Unit Size:
Pack of 35
Pack of 6
Description
For compact and organized sample storage
- Stores vials in clear polystyrene case
- Foam inserts and partitions provide protection
- With alphanumerical indexing card for identification
- Index cards and partitions also available separately
Specifications
Specifications
| For Use With (Application) | Storage and Transportation of Sample Vials |
| For Use With (Equipment) | W228790 Hinged Case |
| Product Type | Partition |
| Quantity | 6 Case |