Learn More
DWK Life Sciences Wheaton™ Leak-Resistant HDPE Bottles with Caps
Product Code. p-3517022 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Neck Finish:
13-425
20-410
24-410
28-410
38-430
Unit Size:
Pack of 24
Pack of 48
Pack of 72
Product Code. Capacity (Metric) Neck Finish unitSize
10613703 4 mL 13-425 Pack of 72
10244732 8 mL 20-410 Pack of 72
10465052 15 mL 20-410 Pack of 72
10729864 30 mL 20-410 Pack of 72
10041452 60 mL 20-410 Pack of 72
10366612 125 mL 24-410 Pack of 72
10284872 250 mL 24-410 Pack of 72
10653703 500 mL 28-410 Pack of 48
10306712 1000 mL 38-430 Pack of 24
9 options
Product Code. 10613703
Brand: DWK Life Sciences 209041
Only null left
Add to basket
Add to basket
With double-seal closure with cap attached
Ideal for short- and long-term storage and packaging
.
  • Narrow mouth
  • No drip pour lip for clean pouring
  • Attached closures remain tight over an extended time
  • Convenience packed
TRUSTED_SUSTAINABILITY
Specifications
Capacity 0.1 oz., 4mL
Capacity (Metric) 4 mL
Capacity (English) 0.1 oz.
Color Natural
Diameter (English) 0.62 in.
Material HDPE
Neck Finish 13-425
Certifications/Compliance ISO 9001
Height (Metric) 38 mm
Height (English) 1.4 in.
Closure Material Polypropylene
Closure Size 13-425 mm
Diameter (Metric) 16 mm
Closure Type Screw Cap