Learn More
DWK Life Sciences Wheaton™ LDPE Dropping Bottles
Product Code. p-39125 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
3 mL
6 mL
7 mL
15 mL
30 mL
Color:
Natural
White
Quantity:
100/Cs.
100/cs
1000/Cs.
1000/cs
Unit Size:
Pack of 100
Pack of 1000
Product Code. 10503144
Brand: DWK Life Sciences W242831A
Only null left
Add to basket
Add to basket
Bottle made of natural low density polyethylene and manufactured to strict quality control standards to ensure the best product for your application.
  • Available in a variety of sizes
  • Packaged in cases of 100, 1000 or bulk
Order Info
Dropping tips or caps must be purchased separately
TRUSTED_SUSTAINABILITY
Specifications
Capacity 3 mL, 0.1 oz.
Capacity (Metric) 3 mL
Capacity (English) 0.1 oz.
Color White
Diameter (English) 0.51 in.
Material LDPE
Quantity 1000/Cs.
Closure Included No
Height (Metric) 39 mm
Product Type Dropping Bottle
Height (English) 1.54 in.
Closure Size 8-425 mm
Diameter (Metric) 14 mm