Learn More
DWK Life Sciences Wheaton™ Lab 45 Graduated Bottles Without Cap or Pour Ring
Product Code. 12432531 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Pack of 12
Pack of 6
Product Code. 12432531
Brand: DWK Life Sciences 219920
Only null left
Add to basket
Add to basket
WHEATON™ media bottles are manufactured from borosilicate glass that conforms to USP Type I requirements and has a low alkali content to help prevent changes in pH to maintain the purity of its contents.
  • Manufactured from USP Type I borosilicate glass helps prevent pH changes
  • 45mm screw thread, wide mouth finish
  • Graduated with writing patch
  • Special pour lip facilitates easy pouring
  • Autoclavable
  • Color coded caps can be purchased separately
TRUSTED_SUSTAINABILITY
Specifications
Capacity 33.8 oz., 1000mL
Capacity (Metric) 1000 mL
Capacity (English) 33.8 oz.
Color Clear
Diameter (English) 3.97 in.
Material Borosilicate Glass
Graduated Yes
Mouth Wide
Height (Metric) 227 mm
Product Type Media Bottle
Height (English) 8.93 in.
Closure Size 45 mm
Diameter (Metric) 101 mm
Closure Type Screw Cap