Learn More
DWK Life Sciences Wheaton™ Lab 45 Graduated Bottles With Cap Attached
Product Code. 12482531 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Pack of 12
Pack of 6
Product Code. 12482531
Brand: DWK Life Sciences 219930
Only null left
Add to basket
Add to basket
Graduated bottles with cap attached
TRUSTED_SUSTAINABILITY
Specifications
Capacity 33.8 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 33.8 oz.
Color Clear
Diameter (English) 3.97 in.
Includes Pouring ring and screw cap
Material Borosilicate Glass
Closure Color White
Graduated Yes
Mouth Wide
Height (Metric) 233 mm
Height (English) 9.17 in.
Closure Material Polypropylene
Closure Size 45 mm
Diameter (Metric) 101 mm
Closure Type Screw Cap