Learn More
DWK Life Sciences Wheaton™ Graduated Valumetric™ Bottles: With White PP Cap With Polyvinyl Liner
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
Quantity:
144/Pk.
24/Pk.
288/Pk.
432/pk.
48/Pk.
96/Pk.
Unit Size:
Pack of 144
Pack of 24
Pack of 288
Pack of 432
Pack of 48
Pack of 96
Description
For storage and samples of solids, dry powdered materials and liquids
- Ideal for use as sample bottles
- Useful for dry powders, soil samples, and specimen storage
- Made from Wheaton 800 glass, which meets USP Type III standards
- Graduated in mL and oz.
Autoclavable black phenolic cap with rubber liner.
Specifications
Specifications
| Capacity (Metric) | 125 mL |
| Capacity (English) | 4 oz. |
| Color | Clear |
| Diameter (English) | 2 in. |
| Material | Glass |
| Quantity | 24/Pk. |
| Closure Color | White |
| Height (Metric) | 104 mm |
| Height (English) | 4 in. |
| Closure Liner | Poly/Vinyl |
| Show More |