Learn More
DWK Life Sciences Wheaton™ Glass Sample Vials with Snap Caps
Product Code. 10712667 Shop All Wheaton Science Products Products
Change view
Click to view available options
Volume (Metric):
16.0 mL
30.0 mL
60.0 mL
120.0 mL
Diameter (Metric) Outer:
30 mm
37 mm
45 mm
50 mm
Height (Metric):
40 mm
50 mm
60 mm
95 mm
Unit Size:
Pack of 144
Pack of 72
Product Code. Volume (Metric) Diameter (Metric) Outer Height (Metric) unitSize
10712667 16.0 mL 30 mm 40 mm Pack of 144
11531225 120.0 mL 50 mm 95 mm Pack of 72
11768699 60.0 mL 45 mm 60 mm Pack of 72
11758699 30.0 mL 37 mm 50 mm Pack of 72
4 options
Product Code. 10712667
Brand: DWK Life Sciences 225536
Only null left
Add to basket
Add to basket
Ideal for collecting, storing, and exhibiting sample in the lab or field, Wheaton™ Glass Sample Vials with Snap Caps contain caps which fit over a special sealing ring to provide effective barrier against moisture.
  • Excellent storage of hygroscopic materials
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Diameter (Metric) Outer 30 mm
Material Soda Lime Glass
Volume (Metric) 16.0 mL
For Use With (Application) For collecting, storing and exhibiting sample in the lab or field
Volume (English) 4.3 dr.
Product Line Wheaton
Volume (Metric) Total 16 mL
Closure Material LDPE
Closure Type Snap Cap
Certifications/Compliance USP Type III
Height (Metric) 40 mm
Product Type Vial