Learn More
DWK Life Sciences Wheaton™ E-Z Vial™ Convenience Pack - Amber Vials with 11mm Crimp Finish
Shop All Wheaton Science Products ProductsDescription
- Manufactured from certified amber Type I borosilicate glass
- Vials are packaged 100 per shelf pack and 10 packs per case
- 40% larger opening improves sample accessibility while reducing autosampler needle damage
Specifications
Specifications
| Color (Seal) | Red |
| Material (Liner) | PTFE/Silicone/PTFE |
| Color | Amber |
| Diameter (Metric) Outer | 12 mm |
| Material | Borosilicate Glass |
| Volume (Metric) | 1.8 mL |
| Height (English) | 1.25 in. |
| Material (Seal) | Aluminum |
| Closure Type | Crimp Cap |
| Quantity | 100 Pk. |
| Show More |