Learn More
DWK Life Sciences Wheaton™ E-Z Vial™ Convenience Pack - Amber Vials with 11mm Crimp Finish
Product Code. 12404901 Shop All Wheaton Science Products Products
Click to view available options
Color (Seal):
Blue
Green
Natural
Red
Quantity:
100 Cs.
100 Pk.
Material (Liner):
PTFE/Rubber
PTFE/Silicone
PTFE/Silicone/PTFE
Unit Size:
Pack of 100
Product Code. 12404901
Brand: DWK Life Sciences W22517201
Only null left
Add to basket
Add to basket
Wheaton E-Z Vials have a 40% larger opening improves sample accessibility while reducing autosampler needle damage.
  • Manufactured from certified amber Type I borosilicate glass
  • Vials are packaged 100 per shelf pack and 10 packs per case
  • 40% larger opening improves sample accessibility while reducing autosampler needle damage
TRUSTED_SUSTAINABILITY
Specifications
Color (Seal) Natural
Material (Liner) PTFE/Rubber
Color Amber
Diameter (Metric) Outer 12 mm
Material Borosilicate Glass
Volume (Metric) 1.8 mL
Height (English) 1.25 in.
Material (Seal) Aluminum
Closure Type Crimp Cap
Height (Metric) 32 mm
Quantity 100 Pk.
Closure Size 11 mm