Learn More
DWK Life Sciences WHEATON™ E-C Sample Vials
Product Code. 10550372 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
1.8 mL
2 mL
2.0 mL
4 mL
4.0 mL
8 mL
8.0 mL
12 mL
16 mL
24 mL
Diameter (Metric) Outer:
12 mm
15 mm
17 mm
19 mm
21 mm
23 mm
Color:
Amber
Clear
Closure Size:
13-425
8-425
Unit Size:
Pack of 200
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Diameter (Metric) Outer Color Closure Size Unit Size
10550372 16 mL 21 mm Clear 13-425 Pack of 200
10343083 4 mL 15 mm Clear 13-425 Pack of 200
13401459 24 mL 23 mm Clear 13-425 Pack of 200
10006621 8.0 mL 17 mm Amber 8-425 Pack of 200
12422741 1.8 mL 12 mm Amber 8-425 Pack of 200
10696833 2.0 mL 12 mm Amber 8-425 Pack of 200
11511185 12 mL 19 mm Clear 13-425 Pack of 200
11728929 8 mL 17 mm Clear 13-425 Pack of 200
11768929 2 mL 12 mm Clear 8-425 Pack of 200
10781323 4.0 mL 15 mm Amber 13-425 Pack of 200
10698052 1.8 mL 12 mm Clear 8-425 Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 10550372 Supplier DWK Life Sciences Supplier No. 224746
  • ONLINE EXCLUSIVE PRICE
    3930.00 NOK
    3174.69 NOK / Pack of 200
    Save 755.31 NOK
    19% Off
View all promotions
Estimated Shipment: 23-02-2027
Economically priced by packaging vials and screw caps separately. WHEATON™ E-C Sample Vials are available in either clear or amber-colored glass.
Economically priced by packaging vials and screw caps separately. Wheaton™ E-C Sample Vials are available in either clear or amber-colored glass.
  • Clear vials made from low extractable borosilicate glass that provides superior chemical resistance
  • Clear vials conform to USP Type I and ASTM E 438 Type I, Class A requirements
  • Amber vials made from borosilicate glass that conforms to USP Type I requirements for light transmission to protect light-sensitive products
  • Vials packed in shrink-wrapped packs of 100 which reduces corrugate contamination
  • 14B rubber lined black phenolic screw caps for aqueous samples
  • PTFE-faced 14B rubber lined black phenolic screw cap for organic samples
  • Vials and caps are autoclavable
Specifications
Material (Liner) PTFE/14B Rubber
Color Clear
Diameter (Metric) Outer 21 mm
Material Borosilicate Glass
Pre-slit No
Sterility Non-sterile
Volume (Metric) 16 mL
Thread Style Screw
Volume (English) 4.3 dr.
Closure Material Phenolic
Closure Type Screw Cap
Quantity 200/Cs.
Closure Size 13-425
Marking Spot No
Autoclavable Autoclavable
Certifications/Compliance ASTM Type I, USP Type I, Class A
Height (Metric) 73 mm
Product Type Sample Vial with Screw Cap