Learn More
DWK Life Sciences Wheaton™ E-C Borosilicate Glass Screw Thread Sample Vials without Caps Product Code.: 11738789
Estimated Shipment: 24-11-2025
Only null left
Add to basket
DWK Life Sciences Wheaton™ E-C Borosilicate Glass Screw Thread Sample Vials without Caps
Product Code. p-205494 Shop All Wheaton Science Products Products
20 mL, Clear, Pack of 200
Click to view available options
Volume (Metric):
1.8 mL
2 mL
2.0 mL
4.0 mL
8.0 mL
12.0 mL
16.0 mL
20 mL
24.0 mL
40 mL
60 mL
Color:
Amber
Clear
Unit Size:
Pack of 143
Pack of 200
Product Code. 11738789
Brand: DWK Life Sciences W224809
Only null left
Estimated Shipment: 24-11-2025
Add to basket
Only null left
Add to basket
Manufactured from Wheaton 33 low extractable borosilicate glass. Wheaton™ E-C Borosilicate Glass Screw Thread Sample Vials without Caps are economically priced and conveniently packed in two shrink-wrapped packs which reduces contamination from corrugated material found in traditional packaging.
  • Caps are packed separately
  • Choice of amber or clear glass
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Diameter (Metric) Outer 28 mm
Material Borosilicate Glass (Low Extractable)
Volume (Metric) 20 mL
Thread Style Screw
Volume (English) 5 dr.
Autoclavable Autoclavable
Closure Type Screw Cap
Height (Metric) 57 mm
Certifications/Compliance ASTM E438, USP Type I, Type I, Class A
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ E-C Borosilicate Glass Screw Thread Sample Vials without Caps >