Learn More
DWK Life Sciences Wheaton™ Connection With Black Phenolic Caps
Product Code. 12475442 Shop All Wheaton Science Products Products
Change view
Click to view available options
Thread Size:
13-425, 20-400
13-425, 22-400
13-425, 24-400
20-400, 24-400
24-400, 24-400
Unit Size:
Pack of 6
Product Code. Thread Size unitSize
12475442 13-425, 24-400 Pack of 6
12477371 13-425, 22-400 Pack of 6
12694773 24-400, 24-400 Pack of 6
12495442 20-400, 24-400 Pack of 6
12465442 13-425, 20-400 Pack of 6
5 options
Product Code. 12475442
Brand: DWK Life Sciences 125496
Only null left
Add to basket
Add to basket
The Wheaton Connection™ Screw Thread Connector Safely joins two exterior threaded glass components without hooks, springs or clamps. Screw cap configuration creates a grease-free, vacuum-tight seal.
  • Inert contact surface
  • Easy to assemble
  • Connection can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
  • Phenolic connectors have a working temperature of up to 180°C
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
Product Type Connector
Thread Size 13-425, 24-400