Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Amber Kits
Product Code. 17992581 Shop All Wheaton Science Products Products
Click to view available options
Closure Color:
Blue
Red
Closure Type:
OmniFlex 3G Igloo Lyo Stopper
OmniFlex 3G Serum Stopper
Ultra Pure Serum Stopper
Volume (Metric):
2 mL
5 mL
10 mL
Unit Size:
1 set
Product Code. 17992581
Brand: DWK Life Sciences W991006CPA
Only null left
Add to basket
Add to basket
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
  • Products supplied ready-to-use
  • Reduces lead times
  • Focuses your efforts on core competencies
  • Eliminates capital investments in equipment needed for cleaning and preparing glassware or plastic containers
  • Frees up valuable clean room space
  • Limits number of suppliers required to provide complete packaging product
  • Amber for photosensitive contents
  • Validated processes
  • Saves valuable time and money while maintaining a high quality product and streamlining process
  • One source for total packaging solutions
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Blue
Color Amber
Material Borosilicate Glass
Pre-slit No
Sterility Sterile
Volume (Metric) 5 mL
For Use With (Application) Light Sensitive Parenteral Preparations and Ready-to-Use Applications
Graduated No
Includes Vial, Stopper, and Seal
No. per Case 120/Cs.
Style Crimp top
Product Line Wheaton CompletePAK
Closure Type Ultra Pure Serum Stopper
Closure Size 20 mm
Marking Spot No
Height (Metric) 40 mm