Learn More
DWK Life Sciences Wheaton™ Complete Cyanide Distillation Apparatus: Components
Product Code. 12481551 Shop All Wheaton Science Products Products
Change view
Click to view available options
Type:
Absorber Top
Cold Finger Condenser
Cold Finger Jacket
Distilling Flask
Inlet Tube
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
12481551 Cold Finger Jacket Each
12471551 Cold Finger Condenser Each
12491551 Distilling Flask Each
12461551 Absorber Top Each
12401561 Inlet Tube Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12481551 Supplier DWK Life Sciences Supplier No. 377164
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
Components for use with Wheaton Complete Cyanide Distillation Apparatus (cat. no. 09108)
Compliance
For use according to ASTM D 2036
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cyanide distillation kit
Type Cold Finger Jacket