Learn More
DWK Life Sciences Wheaton™ Complete Cyanide Distillation Apparatus: Components
Product Code. 12401561 Shop All Wheaton Science Products Products
Change view
Click to view available options
Type:
Absorber Top
Cold Finger Condenser
Cold Finger Jacket
Distilling Flask
Inlet Tube
Unit Size:
Each
Product Code. Type unitSize
12401561 Inlet Tube Each
12481551 Cold Finger Jacket Each
12471551 Cold Finger Condenser Each
12491551 Distilling Flask Each
12461551 Absorber Top Each
5 options
Product Code. 12401561
Brand: DWK Life Sciences 377166
Only null left
Add to basket
Add to basket
Components for use with Wheaton Complete Cyanide Distillation Apparatus (cat. no. 09108)
Compliance
For use according to ASTM D 2036
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cyanide distillation kit
Type Inlet Tube