Learn More
DWK Life Sciences Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined
Shop All Wheaton Science Products Products
Click to view available options
Color:
Green
Natural
Closure Size:
13 mm
20 mm
30 mm
Unit Size:
Pack of 1000
Description
- Good barrier properties
- Color selection includes natural, red, blue and green
- Chemically resistant to alcohol and acetone, among others
Specifications
Specifications
| Color | Green |
| Material | Aluminum |
| Quantity | 1000/Cs. |
| Type | Center Disc Tear-out Cap |
| Closure Size | 13 mm |