Learn More
DWK Life Sciences Wheaton™ Celstir™ Spinner Double Side Arm Flasks without Water Jacket
Product Code. p-123025 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
25 mL
50 mL
125 mL
250 mL
500 mL
1000 mL
3000 mL
8 L
Includes:
Flask only
Flask, impeller and caps
Unit Size:
Each
Product Code. 10557131
Brand: DWK Life Sciences 356875
Only null left
Add to basket
Add to basket
Provide maximum surface between culture and glass flask
  • PTFE-resin coated glass impeller does not protrude through cap and has bottom dimple for more efficient stirring
  • Silicone rubber-lined cap
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Closure Material Silicone Rubber
Diameter (English) Outer 1.4 in.
Capacity (Metric) 50 mL
Material Borosilicate Glass
Height (English) 5.5 in.
Height (Metric) 14.1 cm
Includes Flask, impeller and caps
Side Arm Joint 15-415
Surface Coating PTFE Resin
Autoclavable Autoclavable
Diameter (Metric) Outer 38 mm
Closure Size 38-430