Learn More
DWK Life Sciences Wheaton™ Caps For Liquid Scintillation Vials
Product Code. 12428762 Shop All Wheaton Science Products Products
Click to view available options
Material (Liner):
Linerless
Metal Foil
Polyethylene Disc
Polyseal™
Material:
Polyethylene
Polypropylene
Urea
Closure Size:
15-425
22-400
Unit Size:
Pack of 1000
Product Code. 12428762
Brand: DWK Life Sciences 241017
Only null left
Add to basket
Add to basket
Use as replacement caps or for vials that are provided without caps
Screw Caps For Liquid Scintillation Vials
  • Screw caps for WHEATON liquid scintillation vials
  • Case of 1000 provides an economical alterative
  • Not autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Description Cap Size: 22-400
Material (Liner) Metal Foil
Color White
Material Urea
Type Screw Cap
Closure Size 22-400
For Use With (Equipment) Liquid Scintillation Vial