Learn More
DWK Life Sciences Wheaton™ Caps For Liquid Scintillation Vials
Product Code. 10458531 Shop All Wheaton Science Products Products
Change view
Click to view available options
Material (Liner):
Linerless
Metal Foil
Polyethylene Disc
Polyseal™
Material:
Polyethylene
Polypropylene
Urea
Closure Size:
15-425
22-400
Unit Size:
Pack of 1000
Product Code. Material (Liner) Material Closure Size unitSize
10458531 Metal Foil Urea 15-425 Pack of 1000
10416322 Polyethylene Disc Urea 22-400 Pack of 1000
10653293 Polyseal™ Urea 22-400 Pack of 1000
12428762 Metal Foil Urea 22-400 Pack of 1000
10042003 Linerless Polyethylene 22-400 Pack of 1000
10290379 Metal Foil Polypropylene 22-400 Pack of 1000
6 options
Product Code. 10458531
Brand: DWK Life Sciences 241009
Only null left
Add to basket
Add to basket
Use as replacement caps or for vials that are provided without caps
Screw Caps For Liquid Scintillation Vials
  • Screw caps for WHEATON liquid scintillation vials
  • Case of 1000 provides an economical alterative
  • Not autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Description Cap Size: 15-425
Material (Liner) Metal Foil
Color White
Material Urea
Closure Size 15-425
For Use With (Equipment) Liquid Scintillation Vial
Product Type Screw Cap