Learn More
DWK Life Sciences Wheaton™ Caps For Liquid Scintillation Vials
Shop All Wheaton Science Products ProductsDescription
Screw Caps For Liquid Scintillation Vials
- Screw caps for WHEATON liquid scintillation vials
- Case of 1000 provides an economical alterative
- Not autoclavable
Specifications
Specifications
| Description | Cap Size: 22-400 |
| Material (Liner) | Polyethylene Disc |
| Color | White |
| Material | Urea |
| Closure Size | 22-400 |
| For Use With (Equipment) | Liquid Scintillation Vial |
| Product Type | Screw Cap |