Learn More
DWK Life Sciences Wheaton™ BOD Bottles Replacement Robotic Stopper
Product Code. p-208030 Shop All Wheaton Science Products Products
Change view
Click to view available options
Shape:
Pennyhead
Robotic
Unit Size:
Pack of 12
Product Code. Shape unitSize
12423761 Pennyhead Pack of 12
11708879 Robotic Pack of 12
2 options
Product Code. 12423761
Brand: DWK Life Sciences 227672
Only null left
Add to basket
Add to basket
Compatible with 60 and 300mL BOD bottles
  • Glass, closure-style stopper
  • Unlined
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 60 and 300 mL BOD Bottle
Material Glass
Shape Pennyhead