Learn More
DWK Life Sciences Wheaton™ Black Phenolic Screw Caps With PE Poly-Seal Liner
Product Code. 12413881 Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
13-425
18-400
28-400
38-430
No. per Case:
100 Cs.
144 Cs.
Unit Size:
Pack of 100
Pack of 144
Product Code. 12413881
Brand: DWK Life Sciences 240119
Only null left
Add to basket
Add to basket
Unique cone liner design offers superior torque retention and excellent sealing characteristics
  • Poly-Seal liner is manufactured from low density polyethylene
  • Screw Cap
  • Phenolic
  • Color: Black
It is recommended that liner be tested prior to use for leak resistant applications
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Phenolic Resin
No. per Case 100 Cs.
Neck Finish 28-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Bottle
Closure Liner Polyethylene
Closure Type Screw Cap