Learn More
DWK Life Sciences Wheaton™ Black Phenolic Screw Caps with PTFE faced 14B Rubber Liners for E-Z Ex-Traction™ Vials
Shop All Wheaton Science Products Products
Click to view available options
For Use With (Equipment):
10 mL Vial
2 and 4mL Vial
20mL Vial
8mL Vial
Neck Finish:
13-425
15-425
22-400
24-400
No. per Case:
100 Cs.
1000 Cs.
250 Cs.
500 Cs.
Unit Size:
Pack of 100
Pack of 1000
Pack of 250
Pack of 500
Description
Designed to ensure the ultimate in product safety
- Autoclavable
- Liner made of styrene-butadiene rubber faced with PTFE
- Excellent chemical resistance
Specifications
Specifications
| Color | Black |
| Material | Phenolic Resin |
| No. per Case | 250 Cs. |
| Neck Finish | 15-425 |
| Certifications/Compliance | ISO 9001 |
| For Use With (Equipment) | 8mL Vial |
| Closure Liner | PTFE/Rubber |