missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Amber Glass Reagent Bottle with Ground Glass Stopper
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
Description
- Amber borosilicate glass bottle with narrow mouth
- Protects light-sensitive reagents and solutions
- Ground-glass octagon flat stopper provides excellent seal; protects top lip of bottle from dust
Specifications
Capacity | 16.9 oz., 500mL |
Closure Material | Borosilicate Glass |
Diameter (Metric) | 88 mm |
Height (English) | 6.88 in. |
Height (Metric) | 175 mm |
Capacity (Metric) | 500 mL |
Capacity (English) | 16.9 oz. |
Color | Amber |
Diameter (English) | 3.46 in. |
Material | Glass |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'