Learn More
DWK Life Sciences Wheaton™ Amber Boston Rounds without Caps
Product Code. 12339789 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Closure Size:
20-400 mm
22-400 mm
24-400 mm
28-400 mm
33-400 mm
Unit Size:
Pack of 108
Pack of 160
Pack of 288
Pack of 30
Pack of 432
Pack of 60
Product Code. 12339789
Brand: DWK Life Sciences W216870
Only null left
Add to basket
Add to basket
Amber glass protects light-sensitive solutions
  • Type III soda-lime glass
TRUSTED_SUSTAINABILITY
Specifications
Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Amber
Diameter (English) 3.7 in.
Material Soda Lime Glass
Closure Color White
Height (Metric) 206 mm
Height (English) 8.1 in.
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Size 33-400 mm
Diameter (Metric) 94 mm
Closure Type Screw Cap