Learn More
DWK Life Sciences Wheaton™ Perforated Aluminum Capsules
Shop All Wheaton Science Products Products
Click to view available options
Color:
Blue
Green
Natural
Red
Closure Size:
13 mm
20 mm
Unit Size:
Pack of 1000
Description
Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.
- Good barrier properties
- Color selection includes natural, red, blue and green
- Chemically resistant to alcohol and acetone, among many others
Specifications
Specifications
| Color | Green |
| Material | Aluminum |
| Quantity | 1000/Cs. |
| Closure Size | 13 mm |
| Product Type | Complete Tear-off Cap |