Learn More
DWK Life Sciences Wheaton™ Accessories and Replacement Parts for 20-Slide Glass Staining Dish
Product Code. p-89195 Shop All Wheaton Science Products Products
Change view
Click to view available options
Type:
Slide Staining Rack
Slide Staining Rack Handle
Staining Dish
Unit Size:
Pack of 3
Pack of 6
Product Code. Type unitSize
10512003 Slide Staining Rack Handle Pack of 6
10193482 Slide Staining Rack Pack of 3
10603533 Staining Dish Pack of 3
3 options
Product Code. 10512003
Brand: DWK Life Sciences 900205
Only null left
Add to basket
Add to basket
Featuring an open bottom to facilitate rapid immersion and drainage, reducing carryover. Wheaton™ Accessories and Replacement Parts for 20-Slide Glass Staining Dish are manufactured from soda-lime glass.
Featuring an open bottom to facilitate rapid immersion and drainage, reducing carryover. Wheaton™ Accessories and Replacement Parts for 20-Slide Glass Staining Dish are manufactured from soda-lime glass.
  • Replacement dish and cover
  • Approx. inside bottom dimensions: 91 x 71 x 60mm
TRUSTED_SUSTAINABILITY
Specifications
Material Soda Lime Glass
Type Slide Staining Rack Handle
For Use With (Equipment) Glass Staining Dishes