Learn More
DWK Life Sciences WHEATON™ Accessories and Replacement Parts for 20-Slide Glass Staining Dish
Shop All DWK Life Sciences ProductsDescription
Featuring an open bottom to facilitate rapid immersion and drainage, reducing carryover. Wheaton™ Accessories and Replacement Parts for 20-Slide Glass Staining Dish are manufactured from soda-lime glass.
- Replacement parts for the Wheaton 20-slide glass staining dish unit, the standard for manual staining procedures
- Removable glass slide rack has an open bottom to facilitate rapid immersion and drainage, reducing carryover
- Rack holds 10 single slides, 19 slides arranged alternately, or 20 slides back-to-back in standard sizes: 3 x 1 in. (75 x 25 mm), 3 x 1.5 in. (75 x 38 mm), and 3 x 2 in. (75 x 51 mm)
- Manufactured from soda-lime glass
- Approximate inside bottom dimensions: 91mm L x 71mm W x 60mm D
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Material | Soda Lime Glass |
| Type | Glass Slide Rack |
| For Use With (Equipment) | Glass Staining Dish |
| Slide Count | 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back-to-back |