Learn More
DWK Life Sciences Wheaton™ AC Medium Rounds: With White Polypropylene Cap with Polyvinyl Liner
Product Code. 12389889 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
Closure Size:
33-400 mm
38-400 mm
48-400 mm
58-400 mm
70-400 mm
Unit Size:
Pack of 24
Pack of 48
Product Code. 12389889
Brand: DWK Life Sciences W216998
Only null left
Add to basket
Add to basket
Ideal for use as sample bottles
  • Manufactured from Wheaton 800 glass, which meets USP Type III standards
  • Useful for dry powders, soil samples, and specimen storage
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Clear
Diameter (English) 2.44 in.
Material Soda Lime Glass
Closure Color White
Height (Metric) 129 mm
Height (English) 5.07 in.
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Size 58-400 mm
Diameter (Metric) 62 mm
Closure Type Screw Cap