Learn More
DWK Life Sciences Wheaton™ AC Medium Rounds: With White Polypropylene Cap with Polyvinyl Liner
Shop All Wheaton Science Products ProductsDescription
- Manufactured from Wheaton 800 glass, which meets USP Type III standards
- Useful for dry powders, soil samples, and specimen storage
Specifications
Specifications
| Capacity (Metric) | 125 mL |
| Capacity (English) | 4 oz. |
| Color | Clear |
| Diameter (English) | 2 in. |
| Material | Soda Lime Glass |
| Closure Color | White |
| Height (Metric) | 104 mm |
| Height (English) | 4.09 in. |
| Closure Liner | Poly/Vinyl |
| Closure Material | Polypropylene |
| Show More |