Learn More
DWK Life Sciences Wheaton™ 25mm Filtration Assemblies and Accessories
Choose from a wide range of filtration assemblies and accessories to suit your application needs
Brand: DWK Life Sciences 419332
1255.81 NOK valid until 2024-12-31
Use promo code "21903" to get your promotional price.
Additional Details : Weight : 0.02720kg
Specifications
USP Type I, ASTM E 438 Type I, Class A | |
Glass | |
1 Case |
25mm filtration assembly | |
40 to 60 μm | |
Fritted Glass Base |