Learn More
DWK LIFE SCIENCES™ WHEATON™ 22mm Extractor
Product Code. 12411571 Shop All Wheaton Science Products Products
Change view
Click to view available options
Screw Thread Bottom:
24-400
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Screw Thread Bottom Unit Size
12411571 24-400 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12411571 Supplier DWK LIFE SCIENCES™ Supplier No. 415502
  • ONLINE EXCLUSIVE PRICE
    3040.00 NOK
    2199.10 NOK / Pack of 1
    Save 840.90 NOK
    27% Off
Estimated Shipment: 14-04-2027
The WHEATON™ 22mm Extractor enables continuous extraction of analytes from a solid into an organic solvent. Features Clear-Seal™ joints that eliminate the need for hooks, springs and clamps common to standard ground joint designs.
  • Designed for continuous extraction of analytes from a solid into an organic solvent
  • Clear-Seal™ joints eliminate the need for hooks, springs and clamps common to standard ground joint designs
  • Manufactured from WHEATON 33 low extractable borosilicate glass conforming to ASTM E 438 Type I, Class A requirements
Specifications
Product Type Soxhlet Extractor
For Use With (Application) Used for continuous extraction of analytes from a solid into an organic solvent
Certifications/Compliance ASTM E 438
Screw Thread Bottom 24-400
Screw Thread Top 20-400