Learn More
Cytiva Whatman™ Syringe-Type Membrane Holder
Stainless steel membrane holder for use with a standard syringe
Brand:  Cytiva 1980-002
4132.50 NOK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 0.12000kg
 View more versions of this product
Product Code. 11380644
  • 4350.00 NOK / Each
Estimated Shipment: 21-05-2024
Description
  • Designed for the quick and easy clarification, sterilization, and removal of particulates from small volume samples (typically for HPLC applications)
  • holders contain PTFE gaskets and O-rings and allow the membrane to be autoclaved in place without the filter sticking to the holder.
  • Luer lock fittings connect to a standard syringe and offer convenience and ease of use for clarification, sterilization, and removal of particulates from small volumes of liquid
  • Specifications
    Specifications
    Stainless Steel
    25 mm