Learn More
Cytiva Whatman™ Regenerated Cellulose Membranes: Grade RC 55
Product Code. 11373355
Change view
Click to view available options
Diameter (Metric):
25 mm
47 mm
50 mm
100 mm
110 mm
142 mm
Unit Size:
Pack of 100
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
11373355 142 mm Pack of 25
11383355 25 mm Pack of 100
10166770 50 mm Pack of 100
11375394 110 mm Pack of 25
11365394 47 mm Pack of 100
11363355 100 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11373355 Supplier Cytiva Supplier No. 10410229
Consists of pure cellulose without any wetting agents.
  • Spontaneously wetting, very good wet strength
  • Extremely chemically resistant; suitable for aqueous and organic media
  • Hydrophilic
  • Mechanically stable
  • Sterilizable by all methods
TRUSTED_SUSTAINABILITY
Specifications
Material Regenerated Cellulose
Pore Size 0.5 μm
Hydrophobicity Hydrophilic
Thickness (Metric) 75 μm
Diameter (Metric) 142 mm