Learn More
Cytiva Whatman™ Reeve Angel™ TCLP Glass Fiber Filter Paper
For non-critical industrial laboratory filtration applications
Brand:  Cytiva 5925-142
3215.75 NOK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 0.21000kg
Product Code. 12425382
  • 3385.00 NOK / Pack of 100
Estimated Shipment: 30-09-2024
Description
  • Conforms to EPA method 1311, 5W-846
  • Toxicity characteristic leaching procedure (TCLP)
Specifications
Specifications
Glass Fiber
14.2 cm