Learn More
Cytiva Whatman™ Reeve Angel 226 Creped Qualitative Circles
Click to view available options
Format:
Disc
Sheet
Diameter (Metric):
190 mm
200 mm
330 mm
Unit Size:
Pack of 100
Pack of 1000
Dimensions (L x W):
8 x 8 in.
Description
- High filtration speed
- Retains coarse or gelatinous particulates
- Unique crêped gray surface provides optical contrast for white or dark filtrates
Specifications
Specifications
| Description | 226 19 cm 100/pk |
| Format | Disc |
| Particle Retention | 20 to 25 μm |
| Pore Size | 20 to 25 μm |
| Flow Rate | Fast |
| For Use With (Application) | A general qualitative filter with creped surface. High filtration speed, faster than grade 202 and retain coarse particles gelatinous precipitates. |
| Grade | 226 |
| Quantity | 100/Pk. |
| Diameter (Metric) | 190 mm |