Learn More
Cytiva Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.45μm BA85 Protran Circles
Click to view available options
Quantity:
100/Pk.
1000/Pk.
25/Pk.
50/Pk.
Diameter (Metric):
25 mm
82 mm
82.5 mm
132 mm
137 mm
82 cm
Unit Size:
Pack of 100
Pack of 1000
Pack of 25
Pack of 50
Description
- Universal membrane suitable for Western, Southern, and Northern blotting
- Hydrophilic, so no prewetting with methanol is required
- High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
- Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
- Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
- Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa
Specifications
Specifications
| For Use With (Application) | Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection |
| Binding Capacity | High |
| Diameter (Metric) | 137 mm |
| Quantity | 50/Pk. |
| Format | Disc |
| Material | Nitrocellulose |
| Pore Size | 0.45 μm |
Safety and Handling
Signal Word
- Warning
- Flammable solid Category 2
- H228-Flammable solid.