Learn More
Cytiva Whatman™ Polycap™ HD Disposable Capsules: 75 HD
Product Code. p-107367
Click to view available options
Pore Size:
0.45 μm
1 μm
5 μm
10 μm
20 μm
Inlet Style:
FNPT
Hose Barb
MNPT
Stepped Barb
Outlet Style:
FNPT
Hose Barb
MNPT
Stepped Barb
Unit Size:
Each
Pack of 5
Product Code. 11379124
Brand: Cytiva 2712T
Offers high filtration efficiency and excellent filtrate purity due to its materials and methods of manufacture. Whatman™ Polycap™ HD Disposable Capsules fit between gross filters and microporous membrane filters used for final filtration.
  • Integrity testable
  • With inlet vent
  • Filter Media: Monofilament anisotropic polypropylene (MAPP) filter
  • Able to be sterilized by autoclaving with steam or ethylene oxide (EtO)
  • Retention (with air purge), <2mL
  • Max pressure: 4.1 bar (60 psi)
Filtration of aqueous solutions, biologicals, inks and pigments, photographic emulsions, food and beverages, solvents. Offer broad capacity between gross paper filters and microporous membranes.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Description Polycap HD 75 Capsule Filter, 5.0 μm, with FNPT inlet and outlet (5 pcs)
Material (Membrane) Monofilament Anisotropic Polypropylene
Pore Size 5 μm
Filtration Area 805 cm2
For Use With (Application) Filtration of high-debris solutions and prefiltration of aggressive chemical solutions. Offer broad capacity between gross paper filters and microporous membranes.
Housing Material Polypropylene
Inlet Style FNPT
Max. Pressure 4.1 bar (60 psi)
Outlet Style FNPT