Learn More
Cytiva Whatman™ Phenolphthalein Test Paper on Reel
Product Code. 11310684
Click to view available options
pH Range:
8.3 to 10
Unit Size:
Each
Product Code. 11310684
Brand: Cytiva 2600204A
Combines ease-of-use with accuracy and consistency for determination of the neutralization point in weak acid/strong alkali reactions.
The convenience of using indicator papers for the rapid determination of pH values has led to many applications in laboratories and industry
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use
  • Economical
TRUSTED_SUSTAINABILITY
Specifications
Color Indicator Phenolphthalein
pH Range 8.3 to 10
For Use With (Application) For determination of the neutralization point in weak acid/strong alkali reactions
Quantity Each
Type Phenolphthalein Test Paper