Learn More
Cytiva Whatman™ pH Indicators Integral Comparison Strips
Product Code. 10614282
Change view
Click to view available options
pH Range:
1 to 12
1.8 to 3.8
3.8 to 5.5
5.2 to 6.8
6 to 8.1
8 to 9.7
9.5 to 12
Unit Size:
Pack of 200
Product Code. pH Range unitSize
10614282 5.2 to 6.8 Pack of 200
10451602 3.8 to 5.5 Pack of 200
11370684 1 to 12 Pack of 200
10114071 1.8 to 3.8 Pack of 200
11330694 6 to 8.1 Pack of 200
11340694 8 to 9.7 Pack of 200
11914556 9.5 to 12 Pack of 200
7 options
Product Code. 10614282
Brand: Cytiva 2628990
Only null left
Add to basket
Add to basket
Used for rapid determination of pH values
  • Instant pH readings
  • Accurate for wide range of routine pH testing
  • Convenient and portable for field use
TRUSTED_SUSTAINABILITY
Specifications
pH Range 5.2 to 6.8
Format Strip
For Use With (Application) Routine pH testing
Includes pH indicator and test papers
Type pH Indicator Test Strip, Integral Comparison Strip
Dimensions (L x W) 100 x 11 mm