Learn More
Cytiva Whatman™ pH Indicators Integral Comparison Strips
Click to view available options
pH Range:
1 to 12
1.8 to 3.8
3.8 to 5.5
5.2 to 6.8
6 to 8.1
8 to 9.7
9.5 to 12
Unit Size:
Pack of 200
Description
Used for rapid determination of pH values
- Instant pH readings
- Accurate for wide range of routine pH testing
- Convenient and portable for field use
Specifications
Specifications
| pH Range | 3.8 to 5.5 |
| Format | Strip |
| For Use With (Application) | Routine pH testing |
| Includes | Integral color chart |
| Type | pH Indicator Test Strip, Integral Comparison |
| Dimensions (L x W) | 100 x 11 mm |