Learn More
Cytiva Whatman™ Nuclepore Track-Etched Polycarbonate AOX Membranes
Product Code. 11322905
Click to view available options
Diameter (Metric):
25 mm
47 mm
Unit Size:
Pack of 100
Product Code. 11322905
Brand: Cytiva 111137
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
GE’s Nuclepore filters are manufactured from high-quality polycarbonate film, with sharply defined pore sizes and high flow rates for reproducible AOX analysis.
  • High chemical resistance and thermal stability for a wide range of samples
  • Low, consistent ash levels and low tare weights
  • Compatible with ISO 9562 guidelines for determining adsorbable organically bound halogens
  • Smooth flat surface increases particle visibility
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Nucleopore Polycarbonate
Pore Size 0.4 μm
Flow Rate High
For Use With (Application) Epifluorescence microscopy, environmental analysis, cell biology
Hydrophobicity Hydrophilic
Diameter (Metric) 47 mm