Learn More
Cytiva Whatman™ Nuclepore™ Black Polycarbonate Membranes
Product Code. 11348994
Change view
Click to view available options
Pore Size:
0.2 μm
0.4 μm
0.8 μm
Diameter (Metric):
25 mm
47 mm
Unit Size:
Pack of 100
Product Code. Pore Size Diameter (Metric) unitSize
11348994 0.4 μm 25 mm Pack of 100
11312895 0.2 μm 25 mm Pack of 100
10012101 0.8 μm 25 mm Pack of 100
10208242 0.2 μm 47 mm Pack of 100
4 options
Product Code. 11348994
Brand: Cytiva 110657
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Reduces background fluorescence in support of epifluorescence microscopy. Whatman™ Nuclepore™ Black Polycarbonate Membrane Filters improve visibility and help to distinguish microorganisms from other particulates.
  • Smooth, gridded polycarbonate track-etched (PCTE) membrane surface efficiently supports microorganism enumeration in a standard plate count
  • Extremely low nonspecific absorption 
  • Rapid standard plate count results
  • Compatible with the direct epifluorescent microscopy filter technique (DEFT), which allows microorganisms to be counted directly on a filter
  • DEFT can be conducted in under half an hour, sidestepping the standard > 24 hour process of incubating a microbial sample
  • Microscopic membrane filters are made by track-etching (polycarbonate is bombarded with radiation and chemically treated to create sharp, uniform filter pores)
  • Designed to trap microbes efficiently; more effective than other filter media for certain microorganisms
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Polycarbonate
Pore Size 0.4 μm
For Use With (Application) Epifluorescence technique, potable ultrapure water food and dairy wine and clinical electronics
No. per Pack 100Pack
Type Membrane Filter
Diameter (Metric) 25 mm