Learn More
Cytiva Whatman™ GV 025 Series Glass Vacuum Filtration Manifolds, Replacement Parts
Description
- Description: Replacement Part for GV 025 Series Glass Vacuum Filtration Manifolds
- For Use With (Application): Vacuum Filtration
Specifications
Specifications
| Connectors | NS29 Glass Joint |
| For Use With (Equipment) | Manifold Model GV 025/2 |
| Material | Borosilicate Glass |
| For Use With (Application) | Vacuum Filtration |
| Model | GV 025/2/08 |
| Quantity | 1 Ea. |
| Series | GV025 |
| Type | Erlenmeyer Flask |
| Volume (Metric) | 250 mL |